ESD safety shoes, lace-up, HACCP, Anatom, 2626
Supplier: Abeba
2626/46 2626/47 2626/36 2626/37 2626/38 2626/39 2626/40 2626/41 2626/42 2626/43 2626/44 2626/45 2626/48 2626/52 2626/49 2626/50 2626/51
114-2124EA 111 EUR
114-2124 114-2125 114-2114 114-2115 114-2116 114-2117 114-2118 114-2119 114-2120 114-2121 114-2122 114-2123 ABEB2626/48 ABEB2626/52 ABEB2626/49 ABEB2626/50 ABEB2626/51
ESD safety shoes, lace-up, HACCP, Anatom, 2626
Shoes and Boots
ESD safety shoe made from microfibre upper, reflective material, and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
  • Perspiration-absorbent special lining with silver fibre
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Asymmetric heel and Flexzone for a natural bedding and foot movement
  • TPU torsion waist, arch support and high stability
  • Shock-absorbent PU midsole
  • Protective steel toecap (200 J)
  • Washable at 30 °C
Suitable for hygienically demanding areas.
Colour: White
Certifications: EN 61340 (ESD); DGUV 112-191; HACCP; EN ISO 20345:2011; S1, SRC.