Signs, warning signs, indoor and outdoor
Supplier: KROSCHKE
W1526 W13314 W17044
116-1018EA 83.4 EUR
116-1018 116-1020 116-1021
Signs, warning signs, indoor and outdoor
Signs
The warning signs made from self-adhesive PVC foil are intended to attract attention to objects and situations, in order to warn of potential dangers. According to VBG 125 these must be affixed visibly and permanently at all times.
  • Suitable for indoor and outdoor use
  • Visible from a distance of approx. 6 m
Side length: 200 mm
Certifications: VBG 125 (BGV A8)