Double spatulas
Supplier: KARTELL
594 595
231-2411EA 30.6 EUR
231-2411 231-2412
Double spatulas
Spatulas
Polyamide, fibreglass reinforced, red.
  • Good chemical resistance
  • Heat resistance up to +100 °C (up to +180 °C for short periods)
  • Robust, flexible
Certifications: DIN 12890