Spoon spatulas
Supplier: BOCHEM
3470 3471 3472 3473 3474 3475
231-0184EA 8.8 EUR
231-0184 231-0476 231-0477 231-0478 231-0479 231-0185
Spoon spatulas
Spatulas
Stainless steel ¹⁸/₁₀, anti-magnetic. Multipurpose spoon for a wide range of uses.
  • Round handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean