Safety spectacles, Millennia™ Classic
Supplier: HONEYWELL SAFETY
1002783 1000005 1000003 1000002 1000001 1000006 1000009 1000004 1002781 1002782 1006405
111-1228EA 13.9 EUR
111-1228 111-8302 111-8303 111-8304 111-8305 111-8306 111-8308 111-2294 111-0729 111-0730 111-0734
Safety spectacles, Millennia™ Classic
Safety Glasses
Comfortable safety spectacles with excellent optical qualities for complete protection.
  • Straight temples, no adjustments necessary
  • Comfortable, universal nose bridge
  • Easily replaceable lenses
  • Adjustable, break-away Flexicord retainer for added security and convenience
Weight: 32 g
EN 166, EN 170, EN 171, EN 172 (*); 1F
Certifications: EN 166, EN 170, EN 171, EN 172 (*); 1F