Safety spectacles, uvex super G
Supplier: UVEX
9172.265 9172.110 9172.085 9172.210
111-8062EA 36.6 EUR
111-8062 111-8063 111-1221 111-0463
Safety spectacles, uvex super G
Safety Glasses
These ultra-light, high tech polymer safety glasses have soft and flexible arms. The Super G ensures a comfortable and pressure-free fit that prevents fatigue when in use.
  • Extremely thin and highly break resistant lenses
  • High flexibility and high projectile resistance
  • Aerodynamic shape provides maximum prevention of fog build-up
  • Smooth material transition at the ends of the arms
Weight: 18 g
EN 166-168, EN 170; (EN 172: Anti-UV 5-2,5); 1F
Certifications: EN 166-168, EN 170; (EN 172: Anti-UV 5-2.5); 1F