Sampling spoons, sterile, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H369400000 H369420000 H369460000 H369440000 H369420010 H369440010 H369460010
231-0015EA 579 EUR
231-0015 231-2423 231-2425 231-0418 231-0599 231-0600 231-0638
Sampling spoons, sterile, Sterileware®, SP Bel-Art
Spoons
PS, white. Designed to gather small amounts of sample material.
  • FDA grade materials: Safe for food, drug and cosmetic contact
  • Long handle reaches easily into jars, bottles and other confined areas; minimises the potential of personal contact with sampled material
  • Sterilised by gamma irradiation and individually packed
Packaging: Individually packed, lot stamped for accurate lot tracing, and shipped with a certificate of processing for sterilisation.