VWR®, Magnetic stirring bars, triangular
Supplier: VWR Collection
442-0385EA 23.5 EUR
442-0385 442-0386 442-0387 442-0388 442-0389 442-0390 442-0391 442-0392 442-0393
VWR®, Magnetic stirring bars, triangular
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface. Particularly effective for dissolving solids and mixing sediments because of the scraper-like action on the base of the container.
  • High turbulence, even at low speeds
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)