VWR®, Magnetic stirring bars, double ended
Supplier: VWR Collection
442-0474EA 159 EUR
442-0474 442-0475 442-0476 442-0477 442-0478 442-0479 442-0480 442-0481
VWR®, Magnetic stirring bars, double ended
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface. Very good centring, small footprint and high turbulence even at low speeds. Disc diameter 20 mm, rod diameter 8 mm.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Caution: Note: Coloured PTFE coatings are not as inert as pure PTFE.