Multi layer adhesive floor mats for cleanrooms, BioClean Supatac™
Supplier: Nitritex
STB1845 STB3645 STW2645 STW3645 STW1845
115-3050EA 120 EUR
115-3050 115-3051 115-3053 115-3054 115-1359
Multi layer adhesive floor mats for cleanrooms, BioClean Supatac™
Floor Mats Adhesive Floor Mats
Supatac™ Access Mats are made from LDPE with Polyacrylate adhesive and consist of 30 sheets. They prevent contamination and particulate matter from entering controlled and critical working environments. Manufactured with an anti-bacterial finish, they represent the most advanced technology for significantly reducing the level of traffic-borne contaminants.
  • Suitable for ISO Cleanroom Class 5
  • Uniform thickness and width without pinholes and High Durability Standard Adhesive on entire surface
  • 30 sheets: each sheet coated with a unique adhesive
  • Convenient sizes for placement within various locations
  • Numbered tabs on each sheet provide easy identification and separation
Applications: semiconductor and microelectronic manufacturing, pharmaceutical and biotechnology manufacturing, medical device manufacturing, hospital and operating rooms.
Colour: blue or white.
Packaging: Four mats each with 30 numbered layers in a double walled cardboard carton.