Sterile swabs in round tubes
Supplier: DELTALAB
300252 300250 300263 300261 300251 300252DNA
115-8272EA 244 EUR
115-8272 115-8275 300-0334 300-0332 300-0328 300-0329
Sterile swabs in round tubes
Swabs Swabs in Tubes
Sterile dry swabs in impact resistant PP tubes.
  • Sterilised by ethylene oxide
  • Sealing label cap
  • Extra O-ring in the lid
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904