Centrifuge tubes
Supplier: Brand
114817 114818 114820 114821 114822 114823
525-3201EA 364 EUR
525-3201 525-0240 525-0241 525-0242 525-0243 734-1329
Centrifuge tubes
Tubes Centrifuge Tubes
PP, conical, printed graduation, with HDPE flat screw caps.
  • RCF up to 3000×g
  • Screw cap PE with sealing cone
  • Non sterile or gamma irradiated sterile
* Filling volume (volume of work 13 ml).