Culture tubes with screw cap
Supplier: Brand
113941 113943 113935 113931
391-0035EA 89.6 EUR
391-0035 391-0160 391-0034 391-0033
Culture tubes with screw cap
Tubes Culture Tubes
AR® glass, PP screw cap and white elastomer seal.
  • Autoclavable (+121 °C)
  • Wall thickness 1 mm
  • GL thread