Microcentrifuge tubes with attached screw caps, 0,5/1,5/2,0 ml
Supplier: Brand
780700 780701 780702 780703 780704
211-3201EA 142 EUR
211-3201 211-3202 211-3203 211-3204 211-3205
Microcentrifuge tubes with attached screw caps, 0,5/1,5/2,0 ml
Tubes Microcentrifuge Tubes
PP non graduated, PE screw cap with sealing cone.
  • Can withstand centrifugation to 17000×g
  • Ideal for storage of serum and blood samples, also for boiling and centrifugation
  • Positive sealing screw cap with sealing cone
  • Fast opening and closing with 1¼ turn of cap
  • Operating range: −90 to +100 °C
Ordering information: Coloured cap inserts for identification are available separately.