Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Supplier: Simport Scientific
T332-2 T332-2S T332-4 T332-4S T332-5 T332-5S T332-6 T332-6S T332-6SPR T332-7 T332-7S T332-2SPR T332-4SPR T332-5SPR T332-7SPR
720-0508EA 320 EUR
720-0508 212-9568 212-9570 212-9571 212-9573 720-0509 720-0510 212-9575 720-0511 212-9576 720-0512 212-9569 212-9572 212-9574 720-0513
Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Tubes Microcentrifuge Tubes
PP, with O-ring seal screw cap and attachment loop, autoclavable.
  • The O-ring secured in the top of the cap ensures a positive leakproof seal, time after time, keeping the integrity of small samples under even the most adverse conditions
  • Sterile tubes are gamma irradiated and available either plain or with printed graduations and white marking area for sample identification
  • Can be autoclaved to +121 °C in upright position with caps loosened
  • Certified RNase-, DNase-, pyrogen- and DNA-free
Ideal for cryogenic work as well as freezer storage, boiling applications, centrifugation etc. and will fit most standard microcentrifuge rotors. The cap's high profile facilitates manipulation especially in aseptic procedures and can remain attached to the tube in order to prevent potential mix-up and contamination.
Max. RCF: non skirted tubes can be centrifuged up to 20000×g
Temperature range: –196 to +121 °C
Packaging: Sterile tubes are packaged in tamper-proof resealable bags.