Laboratory tubing, Versilon™ SE 200
Supplier: Saint Gobain Life Sciences
AJD00002 AJD00007 AJD00017 AJD00028
VERNAJD00002EA 242 EUR
VERNAJD00002 VERNAJD00007 VERNAJD00017 VERNAJD00028
Laboratory tubing, Versilon™ SE 200
Tubing Transfer Tubing
Versilon™ tubing, crystal clear.
  • Better flexibility than typical rigid fluoropolymer tubings
  • Chemically resistant and inert
  • Non-contaminating fluid path
Comprises two parts: FEP tubing core and Tygon tubing sheath. This combines the flexibility and clearness of the Tygon R 3603 tubing with the chemical resistance of fluoropolymers(FEP). The tubing core is inert, i.e the delivered liquids can be neither extracted nor contaminated. There is no residual smell or taste from contact between the tubing and the substance flowing through it. Therefore, ideally suitable for foodstuffs and drinks.
Temperature resistant from –40 to +77 °C.
Hardness (Shore durometer A): 67
Density: 1,45
Certifications: Complies with the requirements of the FDA for foodstuffs and the USP class IV criteria for biocompatibility.