Tubing, rubber, vacuum
Supplier: VACUUBRAND
20686000 20686001 20686002 20686005
189-3110EA 20 EUR
189-3110 189-3111 189-3112 189-3114
Tubing, rubber, vacuum
Tubing Transfer Tubing
Rubber.
  • Thick walled, inherently stable and flexible rubber tubing
  • Provides a good seal with hose nipples and hose nozzles
  • Can be ordered in full metre lengths