Micro packaging vials with thread, Nalgene®
Supplier: Thermo Fisher Scientific
342805-0020 362800-0005 342800-0005 342800-0020 342805-0005 362805-0005 362800-0020 342800-0015 342800-0045 362805-0020 342810-0005 342810-0020
215-1187EA 188 EUR
215-1187 215-1259 215-1183 215-1184 215-1186 215-1262 215-1261 215-1524 215-1525 215-1629 215-3696 215-3898
Micro packaging vials with thread, Nalgene®
Vials
PPCO, translucent with low metal content. Disposable bottles, excellent as packaging for shipment by airfreight. Also ideal for storing and transporting PCR reagents, diagnostic and pharmaceutical items as well as many other applications. Manufacturing tolerances also permit use with automatic filling lines. Non cytotoxic and non pyrogenic.
  • Can be centrifuged up to 13000×g at room temperature
  • A wide range of different screw caps (e.g. 215-1630) are optionally available
  • Available non sterile or sterile, clear or amber
* fits most biotechnology and diagnostic equipment.
** e-beam irradiation.
Certifications: Comply with the requirements of the USP Class VI and FDA CFR 21.177.1520. Amber bottles comply with the requirements of the USP XXIV for the transmission of light.