Ring weights for vessels
Supplier: HEATHROW
HS8881A HS8881B HS8881C HS8881D HS8882A HS8882B HS8882D
214-1891EA 51.3 EUR
214-1891 214-1892 214-1893 214-1894 214-1942 214-1943 214-1945
Ring weights for vessels
Weights Flask Weights
Lead core, with blue vinyl coating. Open or closed design. Open design is also suitable for vessels with lateral connections. For stabilising glass or plastic vessels on shakers or in water and oil baths, etc. The weights prevent tipping. The coating prevents the vessels from being scratched or causing damage to other surfaces.
  • Good chemical resistance
  • High temperature resistance
  • Available in various sizes for use with a range of vessel sizes