Economy pH electrodes, InLab® Easy
Supplier: Mettler - Toledo
51343010 51343011
662-2818EA 137 EUR
662-2818 662-2819
Economy pH electrodes, InLab® Easy
Electrodes pH Electrodes
Robust and cost-effective electrodes, simple to operate.
  • Maintenance-free gel electrolyte
  • Ideal for highly saline solutions, soft drinks and beer, and measurements in aquariums
  • Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
pH range: 0 to 14
Temperature range: 0 to 80 °C
Reference system: Ag/AgCl
Ordering information: Supplied with quality certificate. Other electrodes on request.