VWR® AVS TITRINORM® Potassium Chloride Electrolytic Solutions
Supplier: VWR Chemicals
83605.260EA 42.8 EUR
83605.260 83605.290 83606.260 83606.290 83605.180
VWR® AVS TITRINORM® Potassium Chloride Electrolytic Solutions
Electrolytes Electrode Filling Solutions
Potassium chloride 3 Molar solution may also be used as an electrode storage solution.
For full information concerning health and safety data please see the SDS on the website.