Portable balances, PL-S Series
Supplier: Mettler - Toledo
12110000 12110010 12110020 12110030 12110090 12110101 12110105 12110111 12110115 12110680
611-0300EA 0 EUR
611-0300 611-0316 611-0317 611-0318 611-0339 611-2201 611-2203 611-2205 611-2207 611-2226
Portable balances, PL-S Series
Balances and Scales
The PL-S series are simple, compact, light precision balances with an rugged ABS housing and clear LCD. The built-in applications include piece counting, % weighing and dynamic weighing plus the ability to Recall the last weight measured. Below balance weighing is available on all models.
  • Calibration/adjustment with external weights at the touch of a button
  • Optional RS232 for connection to printer or PC are available
  • Round glass draught shields with top access on 1 mg models
  • EU verified versions available
  • Battery or mains operation (PL203-S - mains only)
The PL-S range has two versions.
Standard Model PLXX-S/00
With internal rechargeable battery series PLXX-S/10
Calibration/adjustment with external weights at the touch of a button
Optional RS232 for connecting models to a printer or PC are available
Round glass draught shields with top access on 1 mg models
EU verified versions available
Ordering Information: Versions with RS232C interface(s) and versions with 1st stage OIML verification are also available - please ask for details
Ordering information: Versions with RS232C interface(s) and versions with 1st stage OIML verification are also available - please ask for details.