Precision balances, Excellence XS series
Supplier: Mettler - Toledo
11130174 11130159 11130186 11130171 11130153 11130156 11130158 11130161 11130155 11130164 11130165 11130167 11130168 11130170 11130173 11130176 11130179 11130180 11130182 11130183 11130185 11130188 11130189 11130191 11130192 11130279 11130194 11130270 11130272 11130273 11130275 11130276 11130748 11130750 11130751 11130745 11130747
611-1496EA 3947 EUR
611-1496 611-1507 611-1324 611-1506 611-1347 611-1348 611-2035 611-2036 611-2034 611-2037 611-1494 611-2038 611-1495 611-2039 611-2040 611-2041 611-2042 611-1498 611-2043 611-1499 611-2044 611-2045 611-1500 611-2046 611-1501 611-1505 611-2047 611-1502 611-2059 611-1503 611-2060 611-1504 611-2089 611-2090 611-2091 611-2154 611-2155
Precision balances, Excellence XS series
Balances and Scales
The XS precision balances provide fast and accurate weighing. Draught shields are standard on the small square platform size (see items denoted * in the table). The design also features a large, backlit touch screen that facilitates direct access to all functions and an extended range of applications that include density determinations, formulation, piece counting, statistics and dynamic weighing. Below balance weighing is also available. The units communicate in seven languages and software can be downloaded from the internet.
  • FACT fully automatic temperature controlled internal adjustment
  • DeltaTrac graphically displays the available weighing range
  • Adjustable and detachable display gives flexibility in siting the units
  • Improved connectivity which includes Ethernet and Bluetooth options
  • GLP compliant documentation and settings can be password protected
*Includes draught shield.
Ordering information: OIML certified versions for all models in tables below, together with a full range of accessories for Excellence balances are also available. Please see the website or contact the local Avantor sales office.