Whatman™ TurboBlotter Kits and Refills, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
10416306 10416314 10416326
732-4070EA 301 EUR
732-4070 732-4074 732-4079
Whatman™ TurboBlotter Kits and Refills, Whatman products (Cytiva)
Blotting Apparatus
The TurboBlotter™ system is a rapid, downward blotting device for high-resolution transfer of DNA and RNA. The conventional Northern/Southern transfer stack has been turned upside down in order to take advantage of gravity.
  • Rapid - downward capillary transfer allows for alkaline buffer transfers in one hour and neutral (SSC) transfers in three hours
  • Economical - reusable blotting device requires less buffer and blotting paper, and refill packs are available
  • Compact - has a small footprint and is stackable for multiple, simultaneous transfers
  • Efficient - no heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers
  • Easy to use - simple to set up and works without power or vacuum
The TurboBlotter System is a rapid downward blotting device for high-resolution transfer of DNA and RNA. It provides high-resolution alkaline transfers of DNA in less than one hour, and performs neutral (SSC) transfers of DNA and RNA within three hours. Larger device is stackable for multiple, simultaneous transfers. Reusable and easy to assemble, refills use Whatman Nytran SPC membrane and Whatman 3MM Chr and GB003 or GB005 blotting papers for top performance. The conventional Northern/Southern transfer stack has been turned upside down in order to take advantage of gravity. No heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers.
* Each TurboBlotter™ kit includes: Transfer device, 5 sheets of membrane, 40 sheets of 3MM Chr, 100 sheets of GB004, and 5 wicks of 3 MM Chr.
** TurboBlotter™ refills include: 5 sheets of membrane, 40 sheets of 3MM Chr, 100 sheets of GB004 and 5 wicks of 3 MM Chr.
Ordering information: TurboBlotter™ systems include transfer device, 5 sheets of membrane, 40 sheets of GB002 gel blot paper, 100 sheets of GB004 gel blot paper, and 5 wicks of GB002 gel blot paper. Refills include 5 sheets of membrane, 40 sheets of GB002 gel blot paper, 100 sheets of GB004 gel blot paper, and 5 wicks of GB002 gel blot paper.