Bottle-top dispensers, ceramus®
Supplier: HIRSCHMANN
9312000 9312100 9322000 9322100 9332000 9332100 9342000 9342100 9352000 9352100 9362000 9362100
613-3223EA 416 EUR
613-3223 613-3224 613-3227 613-3228 613-3231 613-3232 613-3236 613-3237 613-3243 613-3244 613-3253 613-3254
Bottle-top dispensers, ceramus®
Bottle Top Dispensers
Fixed or variable volume, autoclavable.
  • Micrometric adjustment knob for fine tuning
  • Ceramic piston made of pure aluminium oxide and cylinder in DURAN® glass guarantee a chemical and thermal inertia
  • Convenient recirculation system for rapid flushing with no loss of reagents
Ordering information: Supplied with A 45 thread, three adapters (A 32, A 38, S 40), one aspiration tube with screw connection, serial number, and quality certificate.