Kjeldahl distillation ramps
Supplier: GERHARDT
10-0037 10-0038 10-0039 10-0043
705-0481EA 0 EUR
705-0481 705-0482 705-0483 705-0487
Kjeldahl distillation ramps
Distillation Systems Kjeldahl Distillation Systems
  • Includes cooling water reservoir
  • KI 9/16: 600 °C, 230V AC, 1800 W
  • KI 12/26: 650 °C, 400 V, 3 phase, 3480 W
Delivery information: Supplied with glassware and fittings.