Accessories for compact automatic extraction units, Soxtherm
Supplier: GERHARDT
13-0062 13-0063 13-0011 1000774 13-0012 13-0051 13-0010 13-0073 13-0047 13-0050 13-0095 13-0096
GERH13-0062EA 37 EUR
GERH13-0062 GERH13-0063 705-0507 GERH1000774 705-0542 701-0074 701-0072 GERH13-0073 GERH13-0047 701-0258 GERH13-0095 GERH13-0096
Accessories for compact automatic extraction units, Soxtherm
Extractors Automatic Extractor Systems
Reliable and effective control including unattended operation of up to 4 units simultaneously can be best carried out using either the compact Multistat controller or with the convenient software Soxtherm Manager which can be operated by most PCs. The software gives an overview of operation and status and provides wide ranging logging functions and associated documentation.
Ordering information: Please ask Avantor for detail of other available accessories including thimbles and seals.