Disinfectant solutions, Bacillol AF
Supplier: BODE CHEMIE
980604 973380 973385 973389 9802174
BODE980604EA 0 EUR
BODE980604 148-0138 148-0137 148-0139 BODE9802174
Disinfectant solutions, Bacillol AF
Cleaning Agents Disinfectants
Alcohol based disinfectant, aldehyde-free and fragrance-free, suitable for spray wipe of all surfaces resistant to alcohol in the agro-food and pharmaceutical industries, in hospital environments or in medical practices. Suitable for application where rapid disinfection is required that leaves no residue.
Contents:
Propan-1-ol 450 mg/g
Propan-2-ol 250 mg/g
Ethanol 47 mg/g
Caution: Use biocides safely. Always read the label and product information before use.
Avantor recommend that you read the package insert and the instructions prior to each use of this product.