Dispersing elements (aggregates) for POLYTRON® PT models
Supplier: KINEMATICA
11030042 11030060 11030030 11030012 11030031 11030062 11030004 11030265 11030299 11030302S
431-0055EA 1913 EUR
431-0055 431-0057 431-0053 431-0051 431-0054 431-0058 431-0142 431-0228 KINE11030299 KINE11030302S
Dispersing elements (aggregates) for POLYTRON® PT models
Homogenisers Dispersing Tools
Designation: EC - EasyClean version for the simplest cleaning or sterilisation (Standard)/2 - rotor/stator with 1 tooth row each.
Ordering information: Other dispersing aggregates available on request.