Dispersing elements for homogenisers
Supplier: IKA
0593400 0594000 1024200 1713300 1713800 2348000 2404000 2466900 2494700 8003000 8003300 8003900 8005100 8006300 L004639 L004640 8003500 8004100 8003200 3304000 3305500 3370100 1684300 4446900 8039500 4446500 4447500 4447300 4446700 4447100 20002971 20002972 20002973 20002974 20002975
431-2453EA 1362 EUR
431-2453 431-2452 431-2451 431-2456 431-2459 431-2455 431-2461 431-2458 431-2211 431-2560 431-2563 431-2569 431-2581 431-2582 431-2602 431-2603 431-0256 431-0257 431-0255 442-0300 442-0310 442-0312 431-0249 431-0253 431-0258 431-0251 431-0223 431-0222 431-0252 431-0254 431-0311 431-0312 IKAA20002973 431-0313 IKAA20002975
Dispersing elements for homogenisers
Homogenisers Dispersing Tools
Type N: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilised by all methods, max. temperature 180 °C.
Type NK: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilised by chemical methods, max. temperature 120 °C.
Type KV: Materials in contact with medium are FFPM/SIC and stainless steel AISI 316L, suitable for vacuum down to 1 mbar, pressure up to 6 bar and for solvents, can be sterilised by chemical methods, max. temperature 220 °C.
SMK jet mixer head: The vertical flow and the high circumferential speed up to 10000 rpm ensure intensive mixing. The head is used for adding gases or liquids, for lump-free suspension of difficult to dissolve powders or for dissolving sedimented, already hardened substances.
Type KD: Materials in contact with the medium are PTFE, FFKM O-ring and stainless steel AISL 316L, suitable for solvents, can be sterilised by all methods, max. temperature 150 °C.
Caution: More dispersing elements are available on request. Please contact Avantor for details.
ST= sawtooth.