Disposable tubes for ULTRA-TURRAX® Tube Drive and ULTRA-TURRAX® Tube Drive control
Supplier: IKA
3703000 3703300 3699500 3699600 3700500 3700600 3703200 3699700 3699800 3701500 3701600 3701700 3703100 20003207 3700701 10013007
431-2892EA 228 EUR
431-2892 431-2893 531-0480 531-0481 531-0477 531-0478 531-0469 531-0482 531-0483 531-0491 531-0492 531-0493 431-0135 IKAA20003207 IKAA3700701 IKAA10013007
Disposable tubes for ULTRA-TURRAX® Tube Drive and ULTRA-TURRAX® Tube Drive control
Homogenisers Homogenisers Bench
ST-20 and ST-50 tubes comprise of tube, stirring device and closure.
  • Ideal for mixing, stirring and extractions
  • Suitable for preparation of soil sample suspensions and solubility testing for pharmaceuticals
  • 20 and 50 ml volume tubes
DT-20 and DT-50 tubes comprise of tube, dispersion device and closure.
Ideal for dispersion, homogenisation and suspensions
Suitable for use in pharmacokinetics, metabolism studies and diagnosis
20 and 50 ml volume tubes
BMT-20-G and BMT-50-G ball-mill tubes comprise of tube, ball (G = glass) and closure.
Ideal for dry milling of dry and brittle samples (e.g. kaolin, gypsum, coloured pigments, tablets)
Suitable for cell maceration and processing of materials mixed with fluids
20 and 50 ml volume tubes
Ordering information: Other tubes are available, including sterile versions (denoted as gamma) and tubes with pierceable membrane (M) closures for dosing and sampling.