Pocket thermo-hygrometers, testo 608
Supplier: Testo
0560.6081 0560.6082
620-3663EA 90 EUR
620-3663 620-1660
Pocket thermo-hygrometers, testo 608
Hygrometers Digital Thermo-Hygrometers
The testo 608-H1 thermo-hygrometer measures humidity, temperature and dew point non-stop. The 608-H2 alarm hygrometer with LED alarm functions for accurate signals when limits are exceeded.
  • Dew point calculation
  • Maximum/minimum display
  • Humidity sensor not affected by condensation
  • LED alarm if limits are exceeded - for 620-1660 only
  • Accuracy ±1,0 digit @ +25 °C
Td = Dew Point
Weight: 168 g
Delivery information: Supplied with a nine volt battery for one year of use, and a factory-calibration protocol (620-1660).