Deuterium lamps
Supplier: LABTECH
AB-400 AC-400 BE-400 BI-400 CE-400 GI-400 GI-401 GY-400 HP-400 HP-405 JV-401 KA-400 KA-401 KN-400 KN-401 LD-400 LI-401 PE-401 SH-400 SH-402 SP-402 VA-400 VA-401 VA-401E WA-400 WA-401 WA-402 WA-404 WA-405 LI-400
554-0806EA 616 EUR
554-0806 554-0807 554-0811 554-0814 554-0816 554-0818 554-0819 554-0820 554-0821 554-0825 554-0827 554-0831 554-0832 554-0833 554-0834 554-0841 554-0843 554-0844 554-0846 554-0847 554-0851 554-0853 554-0854 554-0855 554-0856 554-0857 554-0858 554-0860 554-0861 554-0842
Deuterium lamps
Lights Deuterium Lamps
Listed below are lamps for many common HPLC-UV detectors, along with a selection for UV spectrophotometers. A wide range of lamps for other applications, including UV/VIS spectrophotometers, fluorescent and atomic absorption spectrometers, are available on request. These lamps offer significant savings on OEM prices at the same high quality. All lamps shown here are produced by manufacturers with assured quality standards and are pre-tested and guaranteed as stated below.
  • Guaranteed performance - product enhancements include low static noise, low deviation