Universal refrigerators
Supplier: Liebherr
KP4220 KP2620 K220
471-1087EA 0 EUR
471-1087 471-1027 471-1085
Universal refrigerators
Refrigerators
These are general purpose refrigerators ideal for the storage of non critical products.
  • Electronic temperature regulation, with digital display
  • Off cycle defrost
  • Interchangeable left or right hinged door
Temperature range: 3 to 8 °C
Other models available on request.