VWR®, Accessories for Analogue, Digital and Pulsing Vortex Shakers
Supplier: VWR Collection
444-7047EA 412 EUR
444-7047 444-7048 444-7049 444-7050 444-7051 444-7052 444-7053 444-7058 444-7059 444-7060 444-0208 444-0209 444-0210 444-0211 444-0212 444-0213 444-0214 444-0215 444-0216 444-0217 444-0218 444-0219
VWR®, Accessories for Analogue, Digital and Pulsing Vortex Shakers
Shakers Vortex Shakers
Tube holder kit (444-7047) consists of:
  • Insert retainer
  • Tube holder for 10 tubes of Ø 9 mm, 8 tubes of Ø 13 mm
  • Tube holder for 8 tubes of Ø 14 mm, 8 tubes of Ø 19 mm
  • Tube holder for 8 tubes of Ø 25/29 mm, 5 tubes of Ø 20 mm
  • Two vessel harnesses (example, Erlenmeyer)
  • Foam insert without holes
Attachment and foam inserts for microplates (444-0210):
Set of two foam inserts for microplates (444-0219) and one insert retainer (444-7048)
Insert holder and foam inserts for microtubes (444-0209):
Set of two foam inserts for microtubes (444-0218) and one insert retainer (444-7048)
Single tube holder (444-0217):
Fits all vortex shakers and holds tubes 64 to 114 mm length, minimum Ø 19 mm. Hands-free mixing. Simple assembly, no tools required. Holds 15 or 50 ml centrifuge tubes.
Ampoule tube adapters (444-0212 and 444-0213)
Clamping devices for up to four vials or tubes. Two sizes available:
For vials/tubes 15 to 17 mm Ø
For vials/tubes 10 to 17 mm Ø
Caution: * Can only be used with insert retainer 444-7048
** Can only be used with tube holder 444-0217