Rotating shaker, Roto-Shake Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
SI-1102 SI-1104 SI-1100 SI-1103
444-9150EA 1850 EUR
444-9150 444-9185 SCIISI-1100 444-0062
Rotating shaker, Roto-Shake Genie®
Shakers Rotating/Rocking Shakers
Designed to provide rotating, rocking and 3-D orbital shaking in one compact, durable unit for complete and even soaking of membrane-based blot assays/washings. Accommodates up to two 153×153 mm plastic bags per side, a 305×305 mm non-slip metal rocker tray for conventional rocker/shaker operations, or up to four metal clip plates per side to accommodate similar or different tube sizes for various mixing motions. Expansion kits are available for increased capacity, and different containers. The platform is easy to clean or decontaminate.
  • Magnetic platform and clip plate system - clips can be mounted at any angle, and hold a variety of containers
  • Multiple functions - rocking, rotating and converts quickly into a 3-D orbital shaker with the optional attachment
  • Rocking tray accommodates larger open containers
  • Magnetic strips secure sealed bags for completely uniform soaking and washing
  • Can be used at 0 to 38 °C (95% relative humidity, non-condensing), in an incubator or cold room
Ordering information: Supplied with non-slip tray, four bag mounting strips and one of each of the following clip plates: for 12×10 to 13, 6×15 to 17 and 3×28 to 30 mm Ø tubes.