VWR®, Stirring shafts
Supplier: VWR Collection
441-0253EA 61.4 EUR
441-0253 441-0254 441-0255 441-0256 441-0257 441-0258 441-0259 441-0260 441-0261
VWR®, Stirring shafts
Stirrers Stirring Shafts
PTFE. For use with stirring blades. Chemically inert. The smooth surface reduces contamination and makes cleaning easier. It also prevents the agitation vessels from being scratched.
  • Excellent chemical resistance
  • Temperature resistant from −200 to 280 °C
Ordering information: Stirring blades must be ordered separately.