Overhead stirrer tools, turbine-type impellers
Supplier: Heidolph Instruments GmbH & Co.KG
509-20000-00 509-21000-00
441-1227EA 102 EUR
441-1227 441-1228
Overhead stirrer tools, turbine-type impellers
Stirrers Stirring Tools
Made from stainless steel.
  • For gassing liquids
  • Ideal for stirring liquids with medium to high viscosity
  • Create radial flow