(+)-Cinchonine, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cinchonine, (S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol
M-Clarity™ quality level = MQ100
857270-100G 857270-25G
SIAL857270-100GEA 137 EUR
SIAL857270-100G SIAL857270-25G
(+)-Cinchonine, Sigma-Aldrich®
(+)-Cinchonine
Formula: C₁₉H₂₂N₂O
MW: 294.4 g/mol
Melting Pt: 261…264 °C
Density: 1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064372
CAS Number: 118-10-5
EINECS: 204-234-6
Merck Index: 13,02308