ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Supplier: Roth Carl
Synonyms: Nadide phosphate disodium salt, Triphosphopyridine nucleotide, disodium salt, NADP-Na2, β-NADP-Na2, NADP disodium salt, Nicotinamide-adenine dinucleotide phosphate disodium salt, 1-((2R,3R,4S,5R)-5-((((((((2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium, disodium salt, β-NADH phosphate disodium salt
AE13.1 AE13.2 AE13.3
ROTHAE13.1EA 45.3 EUR
ROTHAE13.1 ROTHAE13.2 ROTHAE13.3
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Formula: C₂₁H₂₆N₇Na₂O₁₇P₃
MW: 787.37 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065390
CAS Number: 24292-60-2
EINECS: 246-129-8