Laboratory glass cleaner, borer 11 UNIVERSAL
Supplier: Borer Chemie
500100.00-F10W 500100.00-K05W 500100.00-K10W
148-5101EA 29.6 EUR
148-5101 148-5102 148-5103
Laboratory glass cleaner, borer 11 UNIVERSAL
Cleaning Agents Detergents and Surface Cleansers
Liquid, mildly alkaline cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory materials in ultrasonic and soaking baths.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • With a broad cleaning spectrum
  • Perfect results even at low concentrations
  • Working solution suitable for multiple use
  • Contains active chlorine as a cleaning booster and is designed to replace chromosulfuric acid
  • EDTA- and NTA-free