(1S)-(-)-β-Pinene, Sigma-Aldrich®

Supplier: Merck

Synonyms: (-)-Pin-2(10)-ene

M-Clarity™ quality level = MQ300

W290300-1KG-K W290300-8KG-K
SIALW290300-1KG-KEA 74.7 EUR
SIALW290300-1KG-K SIALW290300-8KG-K
(1S)-(-)-β-Pinene, Sigma-Aldrich®
(1S)-(-)-β-Pinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 165
Melting Pt: –61 °C
Density: 0.9034 g/cm³ (20 °C)
Flash Pt: 36
MDL Number: MFCD00001345
CAS Number: 18172-67-3
EINECS: 242-060-2
UN: 2319
ADR: 3 III
Merck Index: 12,07600

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR