Size exclusion chromatography columns, Asahipak GF-HQ series
Supplier: Resonac
F7600004 F6710018 F7600001 F7600002
554-1758EA 2240 EUR
554-1758 554-1763 554-1757 554-1756
Size exclusion chromatography columns, Asahipak GF-HQ series
Chromatography Columns
The GF-HQ series are designed for aqueous (GFC) and organic (GPC) size exclusion chromatography. High performance analysis of samples containing hydrophilic and hydrophobic parts, such as surfactants is possible.
  • Polymer-based SEC columns with excellent solvent resistance
  • Supports both aqueous and organic solvent systems
* Estimated value
Order Now
SPECIFICATIONS
Description GF-7M HQ GF-1G 7B GF-310 HQ GF-510 HQ
Environmentally Preferable
Max. pressure 4,5 MPa (45 bar) 7,0 MPa (70 bar) 6,5 MPa (65 bar)
pH range 2 - 9 2 - 9 2 - 9 2 - 9
Temperature limit 4 - 60 °C 4 - 60 °C 4 - 60 °C 4 - 60 °C