Cesium 99.95+% (trace metals basis)
Supplier: Thermo Fisher Scientific
Danger
219190010 219190050
ACRO219190010EA 159 EUR
ACRO219190010 ACRO219190050
Cesium 99.95+% (trace metals basis)
Cesium
Formula: Cs
MW: 132.91
Boiling Pt: 705 °C (760 mmHg)
Melting Pt: 28.5
Density: 1.873 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00134037
CAS Number: 7440-46-2
EINECS: 231-155-4
UN: 1407
ADR: 4.3 I
Merck Index: 12,02051
Order Now
Specification Test Results
Appearance Golden Metal
Total trace metal impurities ≤500 ppm
Cesium (Cs) ≥99.95 % (metals basis)