Sodium, dry stick
Supplier: Thermo Fisher Scientific
Danger
370212500 370210010
ACRO370212500EA 144 EUR
ACRO370212500 ACRO370210010
Sodium, dry stick
Sodium
Formula: Na
Boiling Pt: 883 °C
Melting Pt: 98 °C
Density: 0,9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085307
CAS Number: 7440-23-5
EINECS: 231-132-9
UN: 1428
ADR: 4.3,I
Merck Index: 13,08641
Order Now
Specification Test Results
Appearance Light grey or silver white Sticks
Calcium (Ca) ≤400 ppm
Potassium (K) ≤300 ppm
Sodium (Na) ≥99.8 %