Barium 99+% in mineral oil, rods, Ø ∼ 2 cm

Supplier: Thermo Fisher Scientific
Danger

317861000 317865000
ACRO317861000EA 206 EUR
ACRO317861000 ACRO317865000
Barium 99+% in mineral oil, rods, Ø ∼ 2 cm
Barium
Formula: Ba
MW: 137.33 g/mol
Boiling Pt: 1640 °C
Melting Pt: 725 °C
Density: 3.51
Storage Temperature: Ambient
MDL Number: MFCD00134031
CAS Number: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3 II
Merck Index: 12,00991

Order Now

Specification Test Results

Appearance Grey Rods
Assay ≥99.0 % Ba (incl. Sr)
Strontium (Sr) ≤0.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR