Calcium ≥98% (metals basis), turnings
Supplier: Thermo Fisher Scientific
Danger
10125.09 10125.22 10125.36
10125.09EA 37.5 EUR
10125.09 10125.22 10125.36
Calcium ≥98% (metals basis), turnings
Calcium
Purity: 98% CAS No.: 7440-70-2 Appearance: Metallic Odour: Odourless UN No.: 1401 MDL No.: MFCD00085314
Formula: Ca
Boiling Pt: 1484 °C (1013 hPa)
Melting Pt: 850 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085314
CAS Number: 7440-70-2
EINECS: 213-179-5
UN: 1401
ADR: 4.3,II
Merck Index: 13,01644