Tellurium ≥99.999% (metals basis), lumps
Supplier: Thermo Fisher Scientific
Warning
10362.14 10362.22 10362.36
10362.14EA 76.5 EUR
10362.14 10362.22 10362.36
Tellurium ≥99.999% (metals basis), lumps
Tellurium
Purity: 99.999+% CAS No.: 13494-80-9 Odour: Odourless MDL No.: MFCD00134062
Formula: Te
Boiling Pt: 990 °C (1013 hPa)
Melting Pt: 450 °C
Density: 8.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
ADR: 6.1,III
Merck Index: 12,09273