Copper 99,999 Metals basis, wire, Puratronic® oxygen_free, Ø 0.64 mm (0.025 in)
Supplier: Thermo Fisher Scientific
00098.G2 00098.G6
00098.G2EA 83.7 EUR
00098.G2 00098.G6
Copper 99,999 Metals basis, wire, Puratronic® oxygen_free, Ø 0.64 mm (0.025 in)
Copper
Purity: 99.999% CAS No.: 7440-50-8 Physical Form: ≈2.87g/m Odour: Odourless MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545