Copper ≥99.5% (metals basis), powder -150 mesh
Supplier: Thermo Fisher Scientific
Warning
10160.22 10160.36 10160.A3
10160.22EA 66.6 EUR
10160.22 10160.36 10160.A3
Copper ≥99.5% (metals basis), powder -150 mesh
Copper
Purity: 99.5% CAS No.: 7440-50-8 Molecular Formula: Apparent density 2.6-2.8g/cm3 Appearance: Copper Odour: Odourless UN No.: 3089 MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545